missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF168 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76324AF750
This item is not returnable.
View return policy
Description
RNF168 Polyclonal antibody specifically detects RNF168 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| RNF168 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 750 | |
| E3 ubiquitin-protein ligase RNF168, EC 6.3.2, EC 6.3.2.-, FLJ39749, ring finger protein 168FLJ35794 | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human RNF168. The immunogen is located within amino acids 440 - 490 of RNF168. Amino Acid Squence: KEVDKEQMVPNRQKGSPD | |
| 0.1 mL | |
| Zinc Finger | |
| 165918 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction