missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF165 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561592
This item is not returnable.
View return policy
Description
Immunogen sequence: VVHEIRNYPY PQLHFLALQG LNPSRHTSAV RESYEELLQL EDRLGNVTRG AVQNTIERFT FPHKYKKRRP QDGKGK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Encoded in regions involved in pericentric inversions in patients with bipolar affective disorder. [provided by RefSeq, Jul 2008].
Specifications
| RNF165 | |
| Polyclonal | |
| Unconjugated | |
| RNF165 | |
| 2900024M11Rik; AI427432; Akd2; Ark2c; arkadia 2; Arkadia-like protein 2C; ARKL2; E3 ubiquitin-protein ligase RNF165; G630064H08Rik; Gm96; RGD1560744; RING finger protein 165; RNF111L2; RNF165 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 494470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZSG1 | |
| RNF165 | |
| Recombinant protein corresponding to Human RNF165. Recombinant protein control fragment (Product #RP-98243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction