missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF149 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93343-0.02ml
This item is not returnable.
View return policy
Description
RNF149 Polyclonal antibody specifically detects RNF149 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RNF149 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| DNA polymerase-transactivated protein 2, DNAPTP2, E3 ubiquitin-protein ligase RNF149, EC 6.3.2.-, ring finger protein 149FLJ90504 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 221-400 of human RNF149 (NP_775918.2). FYYIQRFLYTGSQIGSQSHRKETKKVIGQLLLHTVKHGEKGIDVDAENCAVCIENFKVKDIIRILPCKHIFHRICIDPWLLDHRTCPMCKLDVIKALGYWGEPGDVQEMPAPESPPGRDPAANLSLALPDDDGSDDSSPPSASPAESEPQCDPSFKGDAGENTALLEAGRSDSRHGGPIS | |
| 0.02 mL | |
| Cell Biology | |
| 284996 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction