missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF146 Monoclonal Antibody (K16280_8D7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555652
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: NLARESSADG ADSVSAQSGA SVQPLVSSVR PLTSVDGQLT SPATPSPDAS TSLEDSFAHL QLSGDNTAER SHRGEGEEDH ESPSSGRVPA PDTSIEETES DASSDSEDVS AVVAQHSLTQ QRLLVSNANQ
RNF146 protein function is currently unknown.
Specifications
| RNF146 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q5XIK5, Q9NTX7 | |
| RNF146 | |
| Recombinant human RNF146 (immunogen range is 191-320). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K16280_8D7 | |
| Unconjugated | |
| RNF146 | |
| 2610509H23Rik; dactylidin; E3 ubiquitin-protein ligase RNF146; iduna; RING finger protein 146; RING finger protein 146; E3 ubiquitin-protein ligase RNF146; RING-type E3 ubiquitin transferase RNF146; Rnf146 | |
| Mouse | |
| Protein A | |
| RUO | |
| 308051, 81847 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction