missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF144B Monoclonal Antibody (OTI9H10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524997
This item is not returnable.
View return policy
Description
RNF144B Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes UBE2L3 and UBE2L6 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates such as LCMT2, thereby promoting their degradation. Induces apoptosis via a p53/TP53-dependent but caspase-independent mechanism. However, its overexpression also produces a decrease of the ubiquitin-dependent stability of BAX, a pro-apoptotic protein, ultimately leading to protection of cell death; But, it is not an anti-apoptotic protein per se.
Specifications
| RNF144B | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z419 | |
| RNF144B | |
| Human recombinant protein fragment corresponding to amino acids 1-256 of RNF144B produced in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI9H10 | |
| Unconjugated | |
| RNF144B | |
| bA528A10.3; BC025007; E130105P19Rik; E3 ubiquitin-protein ligase RNF144B; IBR domain containing 2; IBR domain-containing protein 2; IBRDC2; p53-inducible RING finger protein; p53RFP; PIR2; RING finger protein 144B; Rnf144b | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 255488 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction