missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF144B/IBRDC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26306-1-AP-20UL
This item is not returnable.
View return policy
Description
E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes UBE2L3 and UBE2L6 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates such as LCMT2, thereby promoting their degradation. Induces apoptosis via a p53/TP53-dependent but caspase-independent mechanism. However, its overexpression also produces a decrease of the ubiquitin-dependent stability of BAX, a pro-apoptotic protein, ultimately leading to protection of cell death; But, it is not an anti-apoptotic protein per se.Specifications
| RNF144B/IBRDC2 | |
| Polyclonal | |
| Unconjugated | |
| RNF144B | |
| IBRDC2, KIAA0161, p53RFP, ring finger protein 144B, RNF144B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 255488 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z419 | |
| RNF144B | |
| RNF144B/IBRDC2 Fusion Protein Ag23679 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction