missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF144A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562099
This item is not returnable.
View return policy
Description
Immunogen sequence: CTLCLKQYVE LLIKEGLETA ISCPDAACPK QGHLQENEIE CMVAAEIMQR YKKLQFEREV LFDPCRTWCP ASTCQAVC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
UBCE7IP4 (Ubiquitin-conjugating enzyme 7- interacting protein 4), also known as RNF144A (RING finger protein 144A), KIAA0161 or RNF144, is a 292 amino acid single-pass membrane protein that contains one RING-type zinc finger and two IBR-type zinc fingers. Functioning as an E3 ubiquitin-protein ligase, UBCE7IP4 accepts ubiquitin (in the form of a thioester) from E2 ubiquitin-conjugating enzymes, such as UBC8 and UBCH7, and transfers that ubiquitin residue to target substrates. Via its RING finger, UBCE7IP4 may play a role in protein-DNA and protein-protein interactions throughout the cell.
Specifications
| RNF144A | |
| Polyclonal | |
| Unconjugated | |
| RNF144A | |
| E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9781 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50876 | |
| RNF144A | |
| Recombinant protein corresponding to Human RNF144A. Recombinant protein control fragment (Product #RP-100981). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction