missing translation for 'onlineSavingsMsg'
Learn More

RNF141 Antibody, Novus Biologicals™

Product Code. 18372409 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.05 mg
missing translation for 'unitSize'
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372409 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18372409 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' H00050862B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RNF141 Polyclonal antibody specifically detects RNF141 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF141
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_057506.2
Gene Alias C3HC4-like zinc finger protein, MGC8715, ring finger protein 141, Zinc finger protein 230, ZNF230ZFP26
Host Species Mouse
Immunogen RNF141 (NP_057506.2, 1 a.a. - 230 a.a.) full-length human protein. MGQQISDQTQLVINKLPEKVAKHVTLVRESGSLTYEEFLGRVAELNDVTAKVASGQEKHLLFEVQPGSDSSAFWKVVVRVVCTKINKSSGIVEASRIMNLYQFIQLYKDITSQAAGVLAQSSTSEEPDENSSSVTSCQASLWMGRVKQLTDEEECCICMDGRADLILPCAHSFCQKCIDKWSDRHRNCPICRLQMTGANESWVVSDAPTEDDMANYILNMADEAGQPHRP
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 50862
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.