missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26368-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A domit negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.Specifications
| RNF14 | |
| Polyclonal | |
| Unconjugated | |
| RNF14 | |
| ARA54, HFB30, HRIHFB2038, ring finger protein 14, RNF14, TRIAD2, Triad2 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9604 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBS8 | |
| RNF14 | |
| RNF14 Fusion Protein Ag23734 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction