missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF14 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148966
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.
Specifications
| RNF14 | |
| Polyclonal | |
| Unconjugated | |
| RNF14 | |
| 2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); androgen receptor-associated protein 54; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; protein Triad2; RING finger protein 14; RNF14; TRIAD2; Triad2 protein; TRIAD2 type I | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 9604 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UBS8 | |
| RNF14 | |
| E. coli-derived recombinant human ARA54, Met127-Gln261. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction