missing translation for 'onlineSavingsMsg'
Learn More

RNF125 Antibody (1D3), Novus Biologicals™

Product Code. 18328559 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328559 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328559 Supplier Novus Biologicals Supplier No. H00054941M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RNF125 Monoclonal antibody specifically detects RNF125 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF125
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 1D3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060301
Gene Alias E3 ubiquitin-protein ligase RNF125, EC 6.3.2.-, FLJ20456, MGC21737, ring finger protein 125TRAC1, T-cell RING activation protein 1, T-cell ring protein identified in activation screen, TRAC-1
Host Species Mouse
Immunogen RNF125 (NP_060301, 143 a.a. ~ 231 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FCQRELYEDSLLDHCITHHRSERRPVFCPLCRLIPDENPSSFSGSLIRHLQVSHTLFYDDFIDFNIIEEALIRRVLDRSLLEYVNHSNT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 54941
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.