missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF122 Polyclonal Antibody

RNF122 Polyclonal Antibody
Brand: Invitrogen PA5121465
This item is not returnable.
View return policy
Description
RNF122 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Specifications
| RNF122 | |
| Polyclonal | |
| Unconjugated | |
| RNF122 | |
| 1110063C11Rik; FLJ12526; MGC126622; RGD1561238; RING finger protein 122; ring finger protein 122 homolog; RN122; RNF122 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79845 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 4.42 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H9V4 | |
| RNF122 | |
| Recombinant Human RING finger protein 122 protein (65-145aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction