missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89571
This item is not returnable.
View return policy
Description
RNF12 Polyclonal specifically detects RNF12 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RNF12 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RLIM | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSESTSARPSRSERNSTEALTEVPPTRGQRRARSRSPDHRRTRARAERSRSPLHPMSEIPRRSHHSISSQTFEHPLVNETEGSSRTRHHVTLRQQISGPELLSRGLFAASGTRNASQGAGSSDTAASGESTGSGQRPPTIVLDL | |
| 0.1 mL | |
| Zinc Finger | |
| 51132 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
| DKFZp686N06224, E3 ubiquitin-protein ligase RLIM, E3 ubiquitin-protein ligase RNF12, EC 6.3.2, EC 6.3.2.-, FLJ25923, FLJ41093, LIM domain-interacting RING finger protein, MGC15161, NY-REN-43, Renal carcinoma antigen NY-REN-43, RING finger LIM domain-binding protein, RING finger protein 12FLJ42887, ring finger protein, LIM domain interacting, ring zinc finger LIM domain binding protein, ring zinc finger protein NY-REN-43antigen, R-LIM, RNF12FLJ45986 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction