missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF114, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00055905-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MAAQQRDCGGAAQLAGPAAEADPLGRFTCPVCLEVYEKPVQVPCGHVFCSACLQECLKPKKPVCGVCRSALAPGVRAVELERQIESTETSCHGCRKNFFLSKIRSHVATCSKYQNYIMEGVKATIKDASLQPRNVPNRYTFPCPYCPEKNFDQEGLVEHCKLFHSTDTKSVVCPICASMPWGDPNYRSANFREHIQRRHRFSYDTFVDYDVDEEDMMNQVLQRSIIDQSpecifications
| RNF114 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_018683.3 | |
| RNF114 | |
| RNF114 (NP_061153.1, 1 a.a. ∼ 228 a.a) full-length human protein. | |
| RUO | |
| 55905 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| ring finger protein 114 | |
| ZNF313 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- RNF114 purified MaxPab mouse polyclonal antibody (B01P)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction