Learn More
Invitrogen™ RNF113B Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5106784
Product Code. 16603225
  • £441.00 / 100µL
Description
Antibody detects endogenous levels of total RNF113B.
RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNF113B
Polyclonal
Unconjugated
RNF113B
bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1
Rabbit
Affinity chromatography
RUO
140432
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8IZP6
RNF113B
A synthesized peptide derived from human RNF113B(Accession Q8IZP6), corresponding to amino acid residues V92-E142.