missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF113B Monoclonal Antibody (2B11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523047
This item is not returnable.
View return policy
Description
Peptide Sequence: MAAPPSPGRT ADQADQVCTF LFKKPGRKGA AGLRKRPACD PEHGESSSSG DEGDTVAQPP RVAPRPRGLH SWQKAAHGDR RGEEAAPESL DVVYRSTRSA.
RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.
Specifications
| RNF113B | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8IZP6 | |
| RNF113B | |
| RNF113B (NP_849192, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 2B11 | |
| Unconjugated | |
| RNF113B | |
| bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 140432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction