missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF112 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25165-1-AP-150UL
This item is not returnable.
View return policy
Description
The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.Specifications
| RNF112 | |
| Polyclonal | |
| Unconjugated | |
| RNF112 | |
| BFP, ring finger protein 112, RNF112, ZNF179 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22671, 7732 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96DY5, Q9ULX5 | |
| RNF112 | |
| RNF112 Fusion Protein Ag18306 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction