missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF111 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106655
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RNF111.
The protein encoded by this gene is a nuclear RING-domain containing E3 ubiquitin ligase. This protein interacts with the transforming growth factor (TGF) -beta/NODAL signaling pathway by promoting the ubiquitination and proteosomal degradation of negative regulators, like SMAD proteins, and thereby enhances TGF-beta target-gene transcription. As a modulator of the nodal signaling cascade, this gene plays a critical role in the induction of mesoderm during embryonic development. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2012].
Specifications
| RNF111 | |
| Polyclonal | |
| Unconjugated | |
| RNF111 | |
| ARK; Arkadia; E3 ubiquitin-protein ligase Arkadia; hRNF111; ring finger 111; RING finger protein 111; RING-type E3 ubiquitin transferase Arkadia; Rnf111 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 300813, 54778, 93836 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZNA4, Q99ML9 | |
| RNF111 | |
| A synthesized peptide derived from human RNF111(Accession Q6ZNA4), corresponding to amino acid residues Y907-R957. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction