missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF11 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98852CL1
This item is not returnable.
View return policy
Description
RNF11 Polyclonal antibody specifically detects RNF11 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| RNF11 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CGI-123, MGC51169, ring finger protein 11, SID1669, Sid1669p | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RNF11. | |
| 0.1 mL | |
| Zinc Finger | |
| 26994 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction