missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16936-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a ring finger motif, which is known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. EST data suggests the existence of multiple alternatively spliced transcript variants, however, their full length nature is not known.Specifications
| RNF10 | |
| Polyclonal | |
| Unconjugated | |
| RNF10 | |
| KIAA0262, RIE2, ring finger protein 10, RNF10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9921 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N5U6 | |
| RNF10 | |
| RNF10 Fusion Protein Ag10433 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction