missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals RND2 Antibody (2D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008153-M01
This item is not returnable.
View return policy
Description
RND2 Monoclonal antibody specifically detects RND2 in Human samples. It is validated for ELISA, ELISA
Specifications
| RND2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 2D12 | |
| ELISA, Sandwich ELISA | |
| ARHN, GTP-binding protein Rho7, ras homolog gene family, member N, Rho family GTPase 2, RHO7, RhoN, rho-related GTP-binding protein Rho7, rho-related GTP-binding protein RhoN | |
| RND2 (AAH18096, 1 a.a. ~ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MWDTSGSSYYDNVRPLAYPDSDAVLICFDISRPETLDSVLKKWQGETQEFCPNAKVVLVGCKLDMRTDLATLRELSKQRLIPVTHEQGTVLAKQVGAVSYVECSSRSSERSVRDVFHVATVASLGRGHRQLRRTDSRRGMQRSAQLSGRPDRGNEGEIHKDRAKSCNLM | |
| 0.1 mg | |
| Signal Transduction | |
| 8153 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction