missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RNaseP Verification Plate, 384-well
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 384 well plate
Unit Size:
Each
Description
- The 384-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with 384-Well Block.
- Confirm the performance of your instrument by running the RNaseP assay.
- Please see your instrument user guide for full instructions on use.
- The 384-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme.
- Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
- The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well.
- To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
Specifications
Specifications
| Format | 384-well Rnase P Verification Plate |
| Content And Storage | Store at –20°C ± 5°C. |
| Calibrated Dye | Proprietary Mix |
| For Use With (Equipment) | ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx |
| Product Type | Rnase P Verification Plate |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 1 x 384 well plate |
| Calibration Type | Instrument Verification Plate |
| PCR Method | qPCR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction