Learn More
RNASEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22577-1-AP-20UL
 View more versions of this product
Product Code. 16851985
  • £135.00 / 20µL
Description
A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.
Specifications
Specifications
RNASEL
Polyclonal
Unconjugated
RNASEL
2 5A dependent ribonuclease, 2 5A dependent RNase, PRCA1, Ribonuclease 4, Ribonuclease L, RNase L, RNASEL, RNS4
Rabbit
Antigen Affinity Chromatography
RUO
24014, 359726, 6041
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q05823, Q05921
Rnasel
RNASEL Fusion Protein Ag17409
20 μL
Primary
Mouse, Rat, Human