missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNaseL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119400
This item is not returnable.
View return policy
Description
RNaseL Polyclonal Antibody for Western Blot, IHC (P)
A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.
Specifications
| RNaseL | |
| Polyclonal | |
| Unconjugated | |
| RNASEL | |
| 2',5'-oligoisoadenylate synthetase-dependent; 2-5A-dependent ribonuclease; 2-5A-dependent RNAase; 2-5A-dependent RNase; E230029I04Rik; I79_009550; interferon-induced 2-5A-dependent RNase; PRCA1; Ribonuclease 4; ribonuclease L; ribonuclease L (2 5-oligoisoadenylate synthetase-dependent); ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent); ribonuclease L (2, 5-oligoisoadenylate synthetase-dependent); ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent); RNase L; Rnasel; RNS4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100048946, 6041 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q05823 | |
| RNASEL | |
| Recombinant protein Ribonuclease L. The antigen corresponds to amino acid range 358-583 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction