missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94719-0.1ml
This item is not returnable.
View return policy
Description
RNASEH2C Polyclonal antibody specifically detects RNASEH2C in Human samples. It is validated for Western Blot
Specifications
| RNASEH2C | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AGS3RNase H2 subunit C, Aicardi-Goutieres syndrome 3 protein, AYP1FLJ20974, MGC22934, ribonuclease H2 subunit C, ribonuclease H2, subunit C, Ribonuclease HI subunit C, RNase H1 small subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-164 of human RNASEH2C (NP_115569.2). MESGDEAAIERHRVHLRSATLRDAVPATLHLLPCEVAVDGPAPVGRFFTPAIRQGPEGLEVSFRGRCLRGEEVAVPPGLVGYVMVTEEKKVSMGKPDPLRDSGTDDQEEEPLERDFDRFIGATANFSRFTLWGLETIPGPDAKVRGALTWPSLAAAIHAQVPED | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84153 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction