missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNaseH2B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559059
This item is not returnable.
View return policy
Description
Immunogen sequence: VVDNVFPNCI LLLKLPGLEK LLHHVTEEKG NPEIDNKKYY KYSKEKTLKW LEKKVNQTVA ALKTNNVNVS SRVQSTAFFS GDQASTDKEE DYIRYAH Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
| RNaseH2B | |
| Polyclonal | |
| Unconjugated | |
| RNASEH2B | |
| AGS2, DLEU8, Aicardi-Goutieres syndrome 2 protein, RNase H2 subunit B, deleted in lymphocytic leukemia 8, ribonuclease H2 subunit B, ribonuclease HI subunit B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TBB1 | |
| RNASEH2B | |
| Recombit protein corresponding to Human RNaseH2B | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction