missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17527-1-AP-150UL
This item is not returnable.
View return policy
Description
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.Specifications
| RNASEH2B | |
| Polyclonal | |
| Unconjugated | |
| RNASEH2B | |
| AGS2, DLEU8, Ribonuclease H2 subunit B, ribonuclease H2, subunit B, Ribonuclease HI subunit B, RNase H2 subunit B, RNASEH2B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361056, 67153, 79621 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TBB1, Q5XI96, Q80ZV0 | |
| Rnaseh2b | |
| RNASEH2B Fusion Protein Ag11189 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction