Learn More
RNASEH2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17527-1-AP-20UL
 View more versions of this product
Product Code. 16857434
  • £135.00 / 20µL
Description
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
Specifications
RNASEH2B
Polyclonal
Unconjugated
RNASEH2B
AGS2, DLEU8, Ribonuclease H2 subunit B, ribonuclease H2, subunit B, Ribonuclease HI subunit B, RNase H2 subunit B, RNASEH2B
Rabbit
Antigen Affinity Chromatography
RUO
361056, 67153, 79621
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5TBB1, Q5XI96, Q80ZV0
Rnaseh2b
RNASEH2B Fusion Protein Ag11189
20 μL
Primary
Human, Rat, Mouse