missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RNASEH2B Monoclonal specifically detects RNASEH2B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | RNASEH2B |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | RM433 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:100-1:200 |
| Formulation | PBS, 50% Glycerol, 1% BSA |
| Gene Alias | ribonuclease H2, subunit B |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to the C-terminus of RNASEH2B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?