missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35504-100ul
This item is not returnable.
View return policy
Description
RNASEH2A Polyclonal antibody specifically detects RNASEH2A in Human,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RNASEH2A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-299 of human RNASEH2A (NP_006388.2).,, Sequence:, MDLSELERDNTGRCRLSSPVPAVCRKEPCVLGVDEAGRGPVLGPMVYAICYCPLPRLADLEALKVADSKTLLESERERLFAKMEDTDFVGWALDVLSPNLISTSMLGRVKYNLNSLSHDTATGLIQYALDQGVNVTQVFVDTVGMPETYQARLQQSFPGIEVTVKAKADALYPVVSAASICAKVARDQAVKKWQFVEKLQDLDTDYGSGYPNDPKTKAWLKEHVEPVFGFPQFVRFSWRTAQTILEKEAEDVIWEDSASENQEGLRKITSYFLNEGSQARPRSSHRYFLERGLESATSL | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 10535 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction