missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32144
This item is not returnable.
View return policy
Description
RNASEH2A Polyclonal specifically detects RNASEH2A in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RNASEH2A | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_006388.2 | |
| RNASEH2A | |
| The immunogen this antibody was made to, maps to a region between residue 250 to 300 of human Ribonuclease H2, Subunit A using the numbering given in entry NP_006388.2 (GeneID 10535). | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 10535 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction