missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2A Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76981MFV610
This item is not returnable.
View return policy
Description
RNASEH2A Polyclonal antibody specifically detects RNASEH2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| RNASEH2A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 610 SE | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human RNAse H2A. The immunogen is located within amino acids 190 - 240 of RNAse H2A. Amino Acid Squence: LDTDYGSGYPNDPKTKA | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10535 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction