missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RNASEH2A Polyclonal antibody specifically detects RNASEH2A in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RNASEH2A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 250 to 300 of human Ribonuclease H2, Subunit A using the numbering given in entry NP_006388.2 (GeneID 10535). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?