missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lyophilized RNase A

Product Code. 10174711
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174711 50 mg 50mg
10546881 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174711 Supplier Macherey-Nagel™ Supplier No. 740505.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Intended primarily for use with NucleoSpin™ and NucleoBond™ kits. Macherey-Nagel™ RNase A is a freeze-dried enzyme sourced from bovine pancreas. It has RNase activity of 120 U/mg. Available in 50 mg and 100 mg packs.

Ribonuclease (RNase) is an enzyme that catalyzes the degradation of RNA into smaller components. RNase A is one of the hardiest enzymes in common laboratory usage. It is specific for single-stranded RNAs, cleaving the 3'-end of unpaired C and U residues, and ultimately forming a 3'-phosphorylated product via a 2',3'-cyclic monophosphate intermediate. It does not require any cofactors for its activity.

  • CAS: 9001-99-4
  • For research purposes only
  • For use, dissolve in water or resuspension buffer
    • Dissolved in water, it can be stored at 4°C for up to 3 months (in smaller aliquots, at -20°C, for longer)
    • Dissolved in resuspension buffer A1 / S1 / S1-EF / RES / RESEF / RES-BAC, it can be stored at 4°C for up to 6 months

Specifications

Color White
For Use With (Equipment) NucleoSpin™ and NucleoBond™ kits
Storage Buffer A1 / S1 / S1-EF / RES / RESEF
/ RES-BAC
Content And Storage Store at room temperature (18–25°C) for at least 1 year
Source Bovine pancreas
Quantity 50 mg
Product Type Ribonuclease A
Form Freeze-dried (Lyophilized)
compliance-icons
Product Identifier
  • RNase A
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.