missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNASE4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121640
This item is not returnable.
View return policy
Description
RNase 4 and RNase 5/ang 1 are unique among the RNase A ribonuclease genes in that they maintain a complex gene locus that is conserved across species with transcription initiated from tissue-specific dual promoters followed by differential exon splicing. Rnase4 (ribonuclease, RNase A family 4) gene can produce 2 transcripts both encoding 148 amino acid proteins. Rnase 4 is a member of the pancreatic-type of secretory ribonucleases, a subset of the ribonuclease A superfamily. RNase 4 prefers poly(C) as a substrate and hydrolyzes 2',3'-cyclic nucleotides, with a pH optimum near 8.0. mRNA encoding RNase 4 is detectable in human pancreas, lung, skeletal muscle, heart, kidney and placenta; liver represents the most abundant source.
Specifications
| RNASE4 | |
| Polyclonal | |
| Unconjugated | |
| RNASE4 | |
| C730049F20Rik; Rab1; Ribonuclease 4; ribonuclease A B1; ribonuclease A family member 4; ribonuclease, RNase A family 4; ribonuclease, RNase A family, 4; RL3; RNase 4; RNASE4; RNS4 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 56759, 6038 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O55004, P34096 | |
| RNASE4 | |
| Recombinant RNASE4 (Asp30-Gly147) expressed in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction