missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNASE2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119398

 View more versions of this product

Product Code. 17730804

  • £486.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RNASE2 Polyclonal Antibody for Western Blot, IHC (P)

This is a non-secretory ribonuclease. It is a pyrimidine specific nuclease with a slight preference for U. Cytotoxin and helminthotoxin. Selectively chemotactic for dendritic cells. Possesses a wide variety of biological activities.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNASE2
Polyclonal
Unconjugated
RNASE2
EDN; eosinophil-derived neurotoxin; Non-secretory ribonuclease; RAF3; Ribonuclease 2; ribonuclease A F3; ribonuclease A family member 2; ribonuclease US; ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin); RNase 2; RNase UpI-2; RNASE2; RNS2
Rabbit
Antigen affinity chromatography, Protein A
RUO
474169
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
0
RNASE2
Recombinant protein Ribonuclease A2.
200 μL
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.