missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNASE13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102340
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RNASE13.
Does not exhibit any ribonuclease activity. [UniProt].
Specifications
| RNASE13 | |
| Polyclonal | |
| Unconjugated | |
| RNASE13 | |
| epididymis secretory sperm binding protein Li 86p; HEL-S-86p; pancreatic ribonuclease; probable inactive ribonuclease-like protein 13; RAL1; ribonuclease 13; ribonuclease A family member 13 (inactive); ribonuclease A L1; ribonuclease, RNase A family, 13 (non-active); ribonuclease-like protein 13; Rnase13 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 440163, 497071, 497194 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5GAL7, Q5GAM7, Q5GAN3 | |
| RNASE13 | |
| A synthesized peptide derived from human RNASE13(Accession Q5GAN3), corresponding to amino acid residues C68-Q118. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction