missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE11 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | RNASE11 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18385966
|
Novus Biologicals
NBP3-04501-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18361906
|
Novus Biologicals
NBP3-04501-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNASE11 Polyclonal specifically detects RNASE11 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RNASE11 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 122651 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| C14orf6, chromosome 14 open reading frame 6, EC 3.1.27.-, probable ribonuclease 11, ribonuclease 11, ribonuclease, RNase A family, 11 (non-active), RNase 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human RNASE11 (NP_660293.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title