missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNase11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532063
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, U87-MG, SK-N-SH. Predicted reactivity: Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.
Specifications
| RNase11 | |
| Polyclonal | |
| Unconjugated | |
| RNASE11 | |
| C14orf6; epididymis secretory sperm binding protein Li 84p; HEL-S-84p; Probable ribonuclease 11; putative inactive ribonuclease 11; RAJ1; ribonuclease 11; ribonuclease A family member 11 (inactive); ribonuclease A j1; ribonuclease, RNase A family, 11 (non-active); RNase 11; RNASE11; UNQ5832/PRO19669 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.79 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8TAA1 | |
| RNASE11 | |
| Recombinant protein encompassing a sequence within the center region of human RNASE11. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction