missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNase1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576865
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.
Specifications
| RNase1 | |
| Polyclonal | |
| Unconjugated | |
| RNASE1 | |
| AI574248; HP-RNase; RAC1; Rib 1; RIB1; Rib-1; Ribonuclease 1; ribonuclease 1, pancreatic; ribonuclease A; ribonuclease A (pancreatic); ribonuclease A C1; ribonuclease A family member 1, pancreatic; ribonuclease BS-1; Ribonuclease pancreatic; ribonuclease, RNase A family, 1 (pancreatic); RNase 1; RNase A; RNase upI-1; RNASE1; RNAseA; RNS1; Seminal ribonuclease; seminal RNase; seminal RNase (aa 47-124); SRN; S-RNase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19752, 6035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00683, P07998 | |
| RNASE1 | |
| Recombinant full length Human RNASE1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction