Learn More
RNase T2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13753-1-AP-20UL
 View more versions of this product
Product Code. 16822134
  • £135.00 / 20µL
Description
RNASET2 is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human maligcies and chromosomal rearrangement, and has been suggested to function as a tumor suppressor protein. Its expression is suppressed in Human T-cell Leukemia Virus type 1 (HTLV-1) infected cells following the binding of the HTLV-1 Tax protein to the RNASET2 promoter. As Adult T-cell leukemia (ATL) is one of the primary diseases caused by HTLV-1 infection, a reduction in the level of RNASET2 by Tax may play a role in ATL development.
Specifications
Specifications
RNase T2
Polyclonal
Unconjugated
RNASET2
Ribonuclease 6, Ribonuclease T2, RNASE6PL, RNASET2
Rabbit
Antigen Affinity Chromatography
RUO
100037283, 8635
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C0HKG5, O00584
RNASET2, Rnaset2a
RNase T2 Fusion Protein Ag4695
20 μL
Primary
Mouse, Human