missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase T1 (1000 U/μL)

RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues.

Brand:  Thermo Scientific™ EN0541

60.33 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Code : 88

Additional Details : Weight : 1.51950kg

 View more versions of this product

Product Code. 10314820

  • £63.50 / 100000 units
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thermo Scientific Rnase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues (see Figure 1 in Supporting Data). It cleaves the phosphodiester bond between the 3'-guanylic residue and the 5'-OH residue of adjacent nucleotides with the formation of corresponding intermediate 2', 3'-cyclic phosphate. The reaction products are 3'-GMP and oligonucleotides with a terminal 3'-GMP.

Rnase T1 does not require metal ions for activity.

Applications

  • Removal of RNA from DNA preparations
  • RNA sequencing
  • Ribonuclease protection assays. Used in conjunction with RNase A
  • Removal of RNA from recombinant protein preparations
  • Determination of the level of RNA transcripts synthesized in vitro from DNA templates containing a "G-less cassette"
Specifications

Specifications

1000 U/μL
RNase
100,000 U
RNase T1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.