missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (2E9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-351AF350
This item is not returnable.
View return policy
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RNase L | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RNASEL | |
| Purified full-length recombinant human RNaseL produced in insect cells. [UniProtNo. Q05823] | |
| Primary | |
| Human, Hamster, Primate, Mouse, Rat | |
| IgG1 κ |
| Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2000000000 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2′-5′-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2′-5′-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| Mouse | |
| 0.1 mL | |
| 6041 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction