missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ RNase A

Product Code. 15889097
Click to view available options
Quantity:
30 mg
Unit Size:
30mg
This item is not returnable. View return policy

Product Code. 15889097

Brand: Macherey-Nagel™ 740505.30

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Lyophilized (freeze-dried) RNase A enzyme intended for use primarily with NucleoSpin™ and NucleoBond™ kits for nucleic acid isolation and purification.

  • Can be dissolved in water or Resuspension Buffer to prepare working solution
  • Compatible with Resuspension Buffers A1 / S1 / S1-EF / RES / RES-EF / RES-BAC
  • Developed, designed, and sold for research purpose only

Specifications

Concentration 90 to <100%
Color White
For Use With (Application) Nucleic acid purification
For Use With (Equipment) NucleoSpin or NucleoBond kits
CAS 9001-99-4
Content And Storage Room temperature (18°C to 25°C)
Source Bovine pancreas
Quantity 30 mg
Biological Activity RNnase activity: 120 U/mg
Form Freeze-dried
compliance-icons
Product Identifier
  • RNase A
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.