missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter RNAClean XP RNA and cDNA Cleanup Reagent

Product Code. 31723141
Change view
Click to view available options
Volume (Metric):
40 mL
450 mL
Unit Size:
40mL
450mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
31723141 40 mL 40mL
31723129 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Refer to SDS
Product Code. 31723141 Supplier Beckman Coulter Supplier No. A63987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Refer to SDS

This RNAClean kit provides a simple, flexible and highly reproducible method.

  • This Kit is for purifying nucleic acid products generated in common enzymatic reactions such as cDNA synthesis and in vitro transcription (IVT) reactions.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type RNA and cDNA
Product Type RNA cand cDNA Cleanup Reagent
Volume (Metric) 40 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.