missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RNA sample loading buffer, no ethidium bromide
Click to view available options
Quantity:
1mL
2mL
Unit Size:
1mL
2mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide.
Specifications
Specifications
| Form | Liquid |
| Quantity | 1mL |
| Storage Requirements | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction