missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNA sample loading buffer, no ethidium bromide

Product Code. 15414419
Change view
Click to view available options
Quantity:
1mL
2mL
Unit Size:
1mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414419 1mL 1mL
15404419 2mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414419 Supplier Thermo Scientific™ Supplier No. J61937.XF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 1mL
Storage Requirements Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.