missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNA Polymerases

Product Code. 31722950 Shop All Thermo Scientific Products
Change view
Click to view available options
Polymerase:
Poly(A) RNA Polymerase
T7 RNA Polymerase
Concentration:
200 U/μL
5 U/μL
Quantity:
20,000 U
800 U
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Polymerase Concentration Quantity Unit Size
31722950 Poly(A) RNA Polymerase 5 U/μL 800 U Each
31722853 T7 RNA Polymerase 200 U/μL 20,000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31722950 Supplier Thermo Scientific™ Supplier No. EP168AF
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 U/μL Poly(A) Polymerase, 160 μL

Store at -20°C.
Polymerase Poly(A) RNA Polymerase
Product Line TheraPure
For Use With (Application) RNA Polyadenylation
Quantity 800 U
Concentration 5 U/μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.