missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A Rabbit anti-Human, Mouse, Clone: 10C7U2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £432.00
Specifications
| Antigen | RNA Polymerase II/POLR2A |
|---|---|
| Clone | 10C7U2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319716
|
Novus Biologicals
NBP3-16910-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18314702
|
Novus Biologicals
NBP3-16910-100UL |
100 μg |
£432.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Mouse samples. It is validated for Western BlotSpecifications
| RNA Polymerase II/POLR2A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5430 | |
| IgG | |
| Affinity purified |
| 10C7U2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| A phospho specific peptide corresponding to residues surrounding S5 of human RNA Polymerase II/POLR2A | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title