missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A, p Ser5 Antibody (CTD 8A7), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00 - £301.00
Specifications
| Antigen | RNA Polymerase II/POLR2A |
|---|---|
| Clone | CTD 8A7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628997
|
Novus Biologicals
NBP2-50243-0.025ml |
0.025 mL |
£149.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651417
|
Novus Biologicals
NBP2-50243 |
0.1 mL |
£301.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
| RNA Polymerase II/POLR2A | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, Epigenetics, Transcription Factors and Regulators | |
| PBS | |
| 5430 | |
| IgM | |
| Thiophilic Chromatography |
| CTD 8A7 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Yeast | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| Peptide: 10 repeats of synthetic peptide YSPTSPS using chemically sythesized phospho-Ser5. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title